D-(-)-Valine ≥98%

855987-25G 855987-5G
220098-231EA 123.85 USD
220098-231 220098-232
D-(-)-Valine ≥98%
D-(-)-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
MDL Number: MFCD00064219
CAS Number: 640-68-6
EINECS: 211-368-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR