Ammonium perrhenate

Danger

316954-25G 316954-5G
220054-625EA 789.23 USD
220054-625 220054-626
Ammonium perrhenate
Ammonium perrhenate
Formula: H₄NO₄Re
MW: 268.24 g/mol
Density: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00012377
CAS Number: 13598-65-7
EINECS: 237-075-6
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR