Sodium triacetoxyborohydride ≥95%

316393-10KG 316393-100G 316393-1KG 316393-25G 316393-25KG
220055-277EA 578.77 USD
220055-277 220055-276 220055-278 220055-279 220055-280
Sodium triacetoxyborohydride ≥95%
Sodium triacetoxyborohydride
Formula: C₆H₁₀BNaO₆
MW: 211.94 g/mol
Melting Pt: 114…118 °C
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 3396
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR