4-Bromo-2-fluorophenol ≥98%

Warning

Synonyms: 1-Bromo-3-fluoro-4-hydroxybenzene

316296-5G 316296-25G
220055-215EA 578.77 USD
220055-215 220055-214
4-Bromo-2-fluorophenol ≥98%
4-Bromo-2-fluorophenol
Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 90 °C (15 mmHg)
Density: 1.74 g/cm³ (20 °C)
MDL Number: MFCD00011722
CAS Number: 2105-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR