Antimony(III) chloride ≥99.0% ACS

Danger

Synonyms: Antimony trichloride

311375-5G 311375-100G
220054-840EA 38.22 USD
220054-840 220054-838
Antimony(III) chloride ≥99.0% ACS
Antimony (III) chloride
Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 223.5 °C (1013 hPa)
Melting Pt: 73 °C
Density: 3.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR