Methyl oleate ≥99%

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

311111-25G 311111-5G
220054-111EA 219.69 USD
220054-111 220054-112
Methyl oleate ≥99%
Methyl oleate
Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: ~ 218.5 °C (1013 hPa)
Melting Pt: -20 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR