2-Methylisophthalonitrile ≥97%

Synonyms: 2-Methyl-1,3-dicyanobenzene, 2-Methyl-1,3-benzenedicarbonitrile, 2-Methylbenzene-1,3-dicarbonitrile, 2,6-Dicyanotoluene

310999-5G 310999-25G
220054-738EA 81.11 USD
220054-738 220054-737
2-Methylisophthalonitrile ≥97%
2-Methylisophthalonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00012285
CAS Number: 2317-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR