Tetramethylammonium tetrahydroborate

Synonyms: Tetramethylammonium tetrahydridoborate, Tetramethylammonium borohydride

310689-10G 310689-50G
220054-707EA 21.85 USD
220054-707 220054-708
Tetramethylammonium tetrahydroborate
Tetramethylammonium tetrahydroborate
Formula: C₄H₁₆BN
MW: 88.99 g/mol
MDL Number: MFCD00011778
CAS Number: 16883-45-7
EINECS: 240-917-5
UN: 3131
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR