Sodium bromide ≥99.0% ACS

310506-100G 310506-500G
220054-680EA 32.31 USD
220054-680 220054-681
Sodium bromide ≥99.0% ACS
Sodium bromide
Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396 °C (1013 hPa)
Melting Pt: 753 °C
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR