Potassium nitrite ≥96.0% ACS

Danger

310484-500G 310484-100G
220054-678EA 315.69 USD
220054-678 220054-677
Potassium nitrite ≥96.0% ACS
Potassium nitrite
Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 387 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 13,07734

Order Now

Specification Test Results

Assay 96.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR