Lithium chloride ≥99% ACS

Warning

310468-100G 310468-2.5KG 310468-500G
220054-668EA 271.38 USD
220054-668 220054-669 220054-670 220054-671
Lithium chloride ≥99% ACS
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR