4-Bromo-2-nitrophenol ≥98%

309877-5G 309877-25G
220054-485EA 44.37 USD
220054-485 220054-484
4-Bromo-2-nitrophenol ≥98%
4-Bromo-2-nitrophenol
Formula: C₆H₄BrNO₃
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 90…94 °C
MDL Number: MFCD00082540
CAS Number: 7693-52-9
EINECS: 231-707-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR