(Methoxymethyl)triphenylphosphonium chloride

Warning

309567-100G 309567-25G
220054-464EA 135.69 USD
220054-464 220054-465
(Methoxymethyl)triphenylphosphonium chloride
(Methoxymethyl)triphenylphosphonium chloride
Formula: C₂₀H₂₀ClOP
MW: 342.8 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00011800
CAS Number: 4009-98-7
EINECS: 223-664-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR