Nickel(II) perchlorate hexahydrate

Danger

309338-5G 309338-500G 309338-100G
220054-093EA 74.31 USD
220054-093 220054-092 220054-091
Nickel(II) perchlorate hexahydrate
Nickel(II) perchlorate hexahydrate
Formula: Cl₂NiO₈·6H₂O
Melting Pt: 140 °C
MDL Number: MFCD00149804
CAS Number: 13520-61-1
EINECS: 237-124-1
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR