Magnesium perchlorate hexahydrate ≥99%

309303-100G 309303-500G
220054-087EA 55.69 USD
220054-087 220054-088
Magnesium perchlorate hexahydrate ≥99%
Magnesium perchlorate hexahydrate
Formula: Cl₂MgO₈·6H₂O
Melting Pt: 185…190 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00149778
CAS Number: 13446-19-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR