Thallium(I) nitrate ≥99.9%

Danger

Synonyms: Thallium nitrate

309230-25G 309230-100G
220054-080EA 152.62 USD
220054-080 220054-079
Thallium(I) nitrate ≥99.9%
Thallium(I) nitrate
Formula: NO₃Tl
MW: 266.39 g/mol
Melting Pt: 206 °C
Density: 1.273 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011280
CAS Number: 10102-45-1
EINECS: 233-273-1
UN: 2727
ADR: 6.1,II
Merck Index: 12,09400

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR