Rhenium pentachloride

Warning

309192-1G 309192-5G
220054-453EA 378.46 USD
220054-453 220054-454
Rhenium pentachloride
Rhenium pentachloride
Formula: Cl₅Re
MDL Number: MFCD00011197
CAS Number: 13596-35-5
EINECS: 237-042-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR