Potassium bromate ≥99.8% ACS

Danger

309087-100G 309087-5G
220054-443EA 76.31 USD
220054-443 220054-445
Potassium bromate ≥99.8% ACS
Potassium bromate
Formula: KBrO₃
Melting Pt: 350 °C
Density: 3.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 12,07779

Order Now

Specification Test Results

Assay 99.8%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR