Tris(2-hydroxyethyl)-1,3,5-triazinetrione

Synonyms: 1,3,5-Tris(2-hydroxyethyl)isocyanuric acid, 1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione, 1,3,5-Tris(2-hydroxyethyl)isocyanurate

309001-50G 309001-250G
220054-437EA 123.57 USD
220054-437 220054-436
Tris(2-hydroxyethyl)-1,3,5-triazinetrione
Tris(2-hydroxyethyl)-1,3,5-triazinetrione
Formula: C₉H₁₅N₃O₆
MW: 261.23 g/mol
Boiling Pt: 527 °C (1013 hPa)
Melting Pt: 133…136 °C
MDL Number: MFCD00003549
CAS Number: 839-90-7
EINECS: 212-660-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR