2-Bromo-4-fluoroanisole ≥96%

Warning

Synonyms: 2-Bromo-4-fluoro-1-methoxybenzene

308919-5G 308919-100G
220054-431EA 26.92 USD
220054-431 220054-298
2-Bromo-4-fluoroanisole ≥96%
2-Bromo-4-fluoroanisole
Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00012014
CAS Number: 452-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR