1-Butyl-3-methylimidazolium methanesulfonate, Sigma-Aldrich®

30881-1KG-F 30881-100G-F
220054-293EA 330.51 USD
220054-293 220054-292
1-Butyl-3-methylimidazolium methanesulfonate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium methanesulphonate
Formula: C₉H₁₈N₂O₃S
MDL Number: MFCD06798173
CAS Number: 342789-81-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR