Potassium molybdate(VI) ≥98%

Warning

Synonyms: Potassium molybdate, Molybdic acid potassium salt, Dipotassium tetraoxomolybdate, Molybdic acid, potassium salt, Potassium molybdenum oxide, di-Potassium molybdate

308390-500G 308390-100G
220054-283EA 552.31 USD
220054-283 220054-282
Potassium molybdate(VI) ≥98%
Potassium molybdate(VI)
Formula: K₂MoO₄
MW: 238.13 g/mol
Melting Pt: 926 °C
Density: 2.91 g/cm³ (18 °C)
MDL Number: MFCD00011370
CAS Number: 13446-49-6
EINECS: 236-599-2
Merck Index: 13,07732

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR