Indium(III) bromide ≥99%

Warning

Synonyms: Indium tribromide

308285-50G 308285-10G
220054-272EA 184.62 USD
220054-272 220054-271
Indium(III) bromide ≥99%
Indium(III) bromide
Formula: Br₃In
MW: 354.53 g/mol
Boiling Pt: 656 °C (1013 hPa)
Melting Pt: 436 °C
Density: 4.77 g/cm³ (25 °C)
MDL Number: MFCD00011057
CAS Number: 13465-09-3
EINECS: 236-692-8
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR