Discandium trioxide

Synonyms: Scandium oxide, Scandium (III) oxide

307874-5G 307874-1G
220054-391EA 139.15 USD
220054-391 220054-390
Discandium trioxide
Discandium trioxide
Formula: O₃Sc₂
MW: 137.91 g/mol
Melting Pt: 1000 °C
Density: 3.86 g/cm³ (20 °C)
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,08392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR