Rhodium(III) chloride

Synonyms: Rhodium(III) chloride, Rhodium trichloride

307866-500MG 307866-2.5G
220054-388EA 749.23 USD
220054-388 220054-387
Rhodium(III) chloride
Rhodium(III) chloride
Formula: Cl₃Rh
MW: 209.26 g/mol
Density: 5.38 g/cm³ (20 °C)
MDL Number: MFCD00011204
CAS Number: 10049-07-7
EINECS: 233-165-4
Merck Index: 12,08355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR