Sodium chromate ≥98%

Danger

307831-100G 307831-500G 307831-5G
220054-382EA 92 USD
220054-382 220054-383 220054-384
Sodium chromate ≥98%
Sodium chromate
Formula: CrNa₂O₄
Melting Pt: 780 °C
Density: 2.698 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003484
CAS Number: 7775-11-3
EINECS: 231-889-5
UN: 3288
ADR: 6.1,II
Merck Index: 12,08745

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR