Iron(II) oxalate dihydrate ≥99%

Warning

Synonyms: Iron (II) ethanedioate dihydrate, Oxalic acid iron (II) salt dihydrate, Ferrous oxalate dihydrate, Iron(II) oxalate hydrate

307726-500G 307726-25G
220053-914EA 98.31 USD
220053-914 220053-913
Iron(II) oxalate dihydrate ≥99%
Iron(II) oxalate dihydrate
Formula: Fe(C₂O₄)·2H₂O
MW: 179.9 g/mol
Melting Pt: 190 °C
Density: 2.28 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150040
CAS Number: 6047-25-2
EINECS: 208-217-4
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR