Iron(III) sulfate hydrate

Warning

307718-12KG 307718-500G 307718-100G 307718-5G
220053-910EA 372.92 USD
220053-910 220053-911 220053-909 220053-912
Iron(III) sulfate hydrate
Iron(III) sulphate hydrate
Formula: Fe₂(SO₄)₃·xH₂O
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR