1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®

Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

30764-100G-F 30764-1KG-F
220054-379EA 188 USD
220054-379 220053-903
1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium chloride
Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
Melting Pt: ~ 80 °C
Density: ~ 1.1 g/cm³ (20 °C)
Flash Pt: 186 °C
MDL Number: MFCD00074843
CAS Number: 65039-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR