Bismuth chloride oxide ≥99%

Warning

Synonyms: Bismuth oxychloride, Bismuth (III) oxychloride, Bismuth(III) chloride oxide

307610-250G 307610-50G
220054-374EA 335.38 USD
220054-374 220054-375
Bismuth chloride oxide ≥99%
Bismuth chloride oxide
Formula: BiClO
MW: 260.43 g/mol
Density: 7.72 g/cm³ (20 °C)
MDL Number: MFCD00010895
CAS Number: 7787-59-9
EINECS: 232-122-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR