Zinc stearate, Technical Grade

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

307564-25G 307564-3KG 307564-1KG
220054-366EA 47.82 USD
220054-366 220054-367 220054-365
Zinc stearate, Technical Grade
Zinc stearate
Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
Melting Pt: 140 °C
Density: 1.095 g/cm³ (20 °C)
Flash Pt: 276 °C
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
EINECS: 209-151-9
Merck Index: 13,10212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR