Zinc sulfate monohydrate

Danger

307491-500G 307491-100G
220053-228EA 131.69 USD
220053-228 220053-227
Zinc sulfate monohydrate
Zinc sulphate monohydrate
Formula: ZnSO₄·1H₂O
MW: 179.45 g/mol
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 13,10213

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR