Copper(II) chloride dihydrate ACS

Warning

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

307483-500G 307483-100G
220053-226EA 169.23 USD
220053-226 220053-225
Copper(II) chloride dihydrate ACS
Copper(II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR