Dehydroacetic acid

Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

30740-100G 30740-500G
220054-356EA 49.75 USD
220054-356 220054-357
Dehydroacetic acid
Dehydroacetic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 108 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR