2-Butoxyethyl acetate ≥99%

Warning

Synonyms: Butylglycol acetate

307289-25ML 307289-1L
220054-344EA 43.08 USD
220054-344 220054-343
2-Butoxyethyl acetate ≥99%
2-Butoxyethyl acetate
Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: -2 °C
Density: 0.9416 g/cm³ (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00009457
CAS Number: 112-07-2
EINECS: 203-933-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR