α-Chloro-2,4-dinitrotoluene ≥99%

Danger

Synonyms: 2,4-Dinitrobenzyl chloride

307262-10G 307262-1G
220054-338EA 181.77 USD
220054-338 220054-339
α-Chloro-2,4-dinitrotoluene ≥99%
α-Chloro-2,4-dinitrotoluene
Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Melting Pt: 34 - 36 °C
Storage Temperature: Ambient
CAS Number: 610-57-1
EINECS: 210-230-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR