4'-Methylacetanilide ≥99%

Synonyms: p-Acetotoluidid, p-Acetotoluidide, N-(4-Methylphenyl)acetamide

307076-10G 307076-100G
220054-333EA 26.31 USD
220054-333 220054-332
4'-Methylacetanilide ≥99%
4'-Methylacetanilide
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 145…148 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008677
CAS Number: 103-89-9
EINECS: 203-155-4
Merck Index: 13,00076

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR