Decylamine

Synonyms: n-Decylamine, Decan-1-amine, 1-Aminodecane

30692-25ML 30692-5ML
220054-064EA 224 USD
220054-064 220054-065
Decylamine
Decylamine
Formula: H₃C(CH₂)₈CH₂NH₂
MW: 157.3 g/mol
Boiling Pt: 221 °C (1013 hPa)
Density: 0.787 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008149
CAS Number: 2016-57-1
EINECS: 217-957-7
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR