2-Bromoisobutyric acid ≥98%

Synonyms: 2-Bromo-2-methylpropanoic acid, α-Bromoisobutyric acid, 2-Bromo-2-methylpropionic acid

306851-100G 306851-25G
220054-056EA 151.85 USD
220054-056 220054-057
2-Bromoisobutyric acid ≥98%
2-Bromoisobutyric acid
Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 198 - 200 °C (1013 hPa)
Melting Pt: 48 °C
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD00004168
CAS Number: 2052-01-9
EINECS: 218-139-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR