2-Fluoro-4-iodoaniline ≥98%

306606-25G 306606-5G
220053-979EA 82.72 USD
220053-979 220053-980
2-Fluoro-4-iodoaniline ≥98%
2-Fluoro-4-iodoaniline
Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 52…54 °C
MDL Number: MFCD00011738
CAS Number: 29632-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR