Methylcyclohexane-[D₁₄]

Synonyms: Methylcyclohexane-[²H₁₄]

306053-1G 306053-5G
220054-225EA 27.85 USD
220054-225 220054-226
Methylcyclohexane-[D₁₄]
Methylcyclohexane-[D₁₄]
Formula: C₇D₁₄
MW: 112.08 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –126 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001498
CAS Number: 10120-28-2
EINECS: 233-325-3
UN: 2296
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR