Sodium hexametaphosphate ≥96%, crystalline +200 mesh, Sigma-Aldrich®

Synonyms: Polyphosphate sodium salt, Sodium polyphosphate, Calgon, Phosphate glass, water soluble, SHMP

305553-5KG 305553-1KG 305553-25G
220054-181EA 155.08 USD
220054-181 220054-179 220054-180
Sodium hexametaphosphate ≥96%, crystalline +200 mesh, Sigma-Aldrich®
Sodium hexametaphosphate
Formula: (NaO₃P)n
Density: 2.4 g/cm³ (20 °C)
MDL Number: MFCD00134118
CAS Number: 68915-31-1
EINECS: 256-779-4
Merck Index: 12,08814

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR