Octadecylamine ≥97%

Danger

Synonyms: Octadecan-1-amine, 1-Octadecylamine, Stearylamine, 1-Aminooctadecane

305391-100G 305391-25G
220054-158EA 601.54 USD
220054-158 220054-159
Octadecylamine ≥97%
Octadecylamine
Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269.51 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 50…52 °C
Density: 0.8618 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008159
CAS Number: 124-30-1
EINECS: 204-695-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR