5-Amino-3-methylisoxazole

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

304271-5G 304271-1G
220053-506EA 120 USD
220053-506 220054-038
5-Amino-3-methylisoxazole
5-Amino-3-methylisoxazole
Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 84 °C
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR