4-Bromo-2-fluoroaniline ≥98%

Warning

304220-10G 304220-50G
220054-029EA 48.89 USD
220054-029 220054-030
4-Bromo-2-fluoroaniline ≥98%
4-Bromo-2-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 75…80 °C (0.3 mmHg)
Melting Pt: 41…42 °C
MDL Number: MFCD00010221
CAS Number: 367-24-8
EINECS: 206-685-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR