2-Methyl-2-cyclopenten-1-one ≥98%

304190-5G 304190-1G
220054-028EA 229.38 USD
220054-028 220054-027
2-Methyl-2-cyclopenten-1-one ≥98%
2-Methyl-2-cyclopenten-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 158 °C (1013 hPa)
Density: 0.9772 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00012275
CAS Number: 1120-73-6
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR