2-Butynoic acid ≥98%

Danger

303666-5G 303666-1G
220053-648EA 142.15 USD
220053-648 220053-647
2-Butynoic acid ≥98%
2-Butynoic acid
Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 78 °C
Density: 0.9641 g/cm³ (20 °C)
MDL Number: MFCD00004363
CAS Number: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR