Propiolaldehyde diethyl acetal ≥97%

Synonyms: Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne

303607-5G 303607-25G 303607-1G
220053-644EA 150.77 USD
220053-644 220053-643 220053-642
Propiolaldehyde diethyl acetal ≥97%
Propiolaldehyde diethyl acetal
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 38 °C (1013 hPa)
Density: 0.8953 g/cm³ (20 °C)
MDL Number: MFCD00009237
CAS Number: 10160-87-9
EINECS: 233-430-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR