2-Amino-5-nitrophenol

Warning

Synonyms: 2-Hydroxy-4-nitroaniline

303585-100G 303585-25G
220053-638EA 74.31 USD
220053-638 220053-639
2-Amino-5-nitrophenol
2-Amino-5-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 207 - 208 °C
Density: 1.54 g/cm³ (20 °C)
MDL Number: MFCD00007692
CAS Number: 121-88-0
EINECS: 204-503-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR