2-Amino-5-bromopyrimidine

303526-5G 303526-1G
220053-636EA 43.51 USD
220053-636 220053-885
2-Amino-5-bromopyrimidine
2-Amino-5-bromopyrimidine
Formula: C₄H₄BrN₃
MW: 174 g/mol
MDL Number: MFCD00012341
CAS Number: 7752-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR